Pipette tray, SP Bel-Art
Supplier: Bel-Art Products, a part of SP
SP Bel-Art
BELAF189400000EA 80.7 EUR
BELAF189400000
Pipette tray, SP Bel-Art
Trays/Dishes/Pots Pipette Trays
This pipette tray made of PS can be used in drawers or on the benchtop for storing pipettes and can also be used for soaking pipettes before cleaning.
  • Scalloped rims hold 16 small or 7 large pipettes
  • Center support holds 12 short pipettes in a perpendicular direction