VWR® Terrific, Broth, powder
Supplier: VWR
VWR®
J869-500GEA 167 EUR
J869-500G
VWR® Terrific, Broth, powder
Media for Microbiology Dehydrated Media for Microbiology
Terrific broth is an enriched medium, ideal for culturing cells for nucleic acid purification and recombinant protein expression.
  • Save time using pre-mixed media
  • Homogenous and high quality powders
Terrific broth is an enriched medium containing high levels of tryptone and yeast extract, which provide amino acids, vitamins and trace elements necessary for high density growth. Glycerol is added as a carbohydrate source, while the potassium phosphate is present to both maintain optimal pH and serve as a source of potassium for cellular metabolism. Glycerol is not supplied with this media.
1 L of prepared media contains 12 g of tryptone, 24 g of yeast extract, 9,4 g of potassium phosphate dibasic, and 2,2 g of potassium phosphate monobasic.