Desiccators, mini vacuum, SP Bel-Art
Supplier: Bel-Art Products, a part of SP
467-2245EA 227 EUR
467-2245
Desiccators, mini vacuum, SP Bel-Art
Desiccators Desiccators, Plastic
Desiccators with top made of clear polycarbonate, for easy viewing of contents, and white polypropylene bottom. The small size of this desiccator allows quick vacuum creation for rapid, repeated degassing of multiple samples.
  • Vacuum rating of 740 mm Hg
  • Stopcock with 2 mm PTFE plug accepts tubing with ¼ to ³/₈" int.Ø
Peripheral flange around the bottom fits in to two snap locks and seals the two halves against a Buna-N O-ring.