3-Methylbenzyl mercaptan
Supplier: Thermo Scientific
Synonyms:
m-Xylene-α-thiol, 3-Methylbenzylmercaptan
TL00469DA
TL00469EA
TL00469.50
MAYBTL00469DAEA
0
EUR
MAYBTL00469DA
MAYBTL00469EA
MAYBTL00469.50
3-Methylbenzyl mercaptan
3-Methylbenzyl mercaptan
Formula:
C₈H₁₀S MW: 138.23 g/mol Boiling Pt: 215…217 °C (1013 hPa) |
MDL Number:
MFCD00022075 CAS Number: 25697-56-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...