Ampicillin sodium salt, Sigma-Aldrich®

Supplier: ROCHE


M-Clarity™ quality level = MQ100

ROCH10835269001EA 345 EUR
ROCH835269 ROCH10835269001
Ampicillin sodium salt, Sigma-Aldrich®
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
MW: 371,39 g/mol
MDL Number: MFCD00064313
CAS Number: 69-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR