Chicken lysozyme (Muramidase) (from egg white)
Supplier: Merck
L4919-1G L4919-500MG L4919-5G
SIALL4919-1GEA 164 EUR
SIALL4919-1G SIALL4919-500MG SIALL4919-5G
Chicken lysozyme (Muramidase) (from egg white)
Enzymes
Lysozyme is abundantly found in animal and plant kingdoms. It is a natural food preservative and also found to be present in specific bacterial cell walls. Lysozyme is usually present in tears, milk, urine and saliva.Enzyme breaks down the cell walls of bacteria; used to prepare spheroplasts.
Lysozyme from chicken egg white has been used in dielectric spectroscopy studies of dynamics of protein, as a control to measure the human lysosyme activity, as a supplement in soaking solution to treat lenses.