Knitted gloves, Nitras® 3400X, partially coated, with knitted cuff
Supplier: AS ARBEITSSCHUTZ
3400X8 3400X9 3400X10
ASAS3400X8EA 26.7 EUR
ASAS3400X8 ASAS3400X9 ASAS3400X10
Knitted gloves, Nitras® 3400X, partially coated, with knitted cuff
Gloves
Work gloves made of 100% cotton jersey, partially coated with nitrile rubber (NBR).
  • High wearing comfort and flexibility
  • Partially coated: Open back of the hand ensures breathability
  • With knitted waistband
Ideal for construction, industry and commerce.
EN 388 4121
Certifikace: EN 388 (4111X); Cat. II
OEKO-Tex certified (STANDARD 100)