Industrial gloves, Nitras® 03410, partially coated, with knitted cuff
Supplier: AS ARBEITSSCHUTZ
034109 0341010 0341111
112-2399EA 259 EUR
112-2399 112-2403 112-2404
Industrial gloves, Nitras® 03410, partially coated, with knitted cuff
Gloves
Knitted gloves made of 100% cotton jersey, partially coated with nitrile rubber.
  • Offers great comfort and flexibility
  • With knitted waistband
  • Very good grip on dry, wet and slippery objects
For use in the automotive industry, in manufacturing or construction.
EN 388 4221
Certifikace: EN 388 (4221X); Cat. II