Nitrile gloves, Nitras® SUPREME NITRILE 8300
Supplier: AS ARBEITSSCHUTZ
8300S 8300M 8300L 8300XL
ASAS8300SEA 36.3 EUR
ASAS8300S ASAS8300M ASAS8300L ASAS8300XL
Nitrile gloves, Nitras® SUPREME NITRILE 8300
Gloves
Disposable nitrile gloves with a wide range of potential applications: in medical practices, labs, printing works, and the pharmaceutical and food industry.
  • Non sterile, with beaded cuffs
  • Powder-free
  • Extra sensitive, very stretchy
  • Excellent tactile sensitivity
  • Secure grip due to surface structure
EN 455; AQL 1.5
Certifikace: EN 374-1 (Type C), EN 374-5 (VIRUS), EN 455; AQL 1,5
Food contact approved