Chemical resistant gloves, Nitras® 3450
Supplier: AS ARBEITSSCHUTZ
34509 345010 34507 34508
112-2608EA 4.2 EUR
112-2608 112-2609 112-2572 112-2607
Chemical resistant gloves, Nitras® 3450
Gloves
Gloves made from a special nitrile blend.
  • Individually packaged in poly bags
  • Flock-lined
For the chemical, printing and petroleum industries.
Double wall thickness: 0.76 mm
Colour: Green
EN 388 (4001), EN 374 (KLA), EN 374-2
Certifikace: EN 388 (4101X), EN 374-1:2016 Type A (AKLMNO), EN 374-5:2016 VIRUS
Food contact approved