Chemical resistant gloves, Nitras® 3460
Supplier: AS ARBEITSSCHUTZ
34607 34608 34609 346010 346011
112-2610EA 6.3 EUR
112-2610 112-2611 112-2612 112-2613 112-2614
Chemical resistant gloves, Nitras® 3460
Gloves
Lightweight chloroprene gloves with structured palm for better grip.
  • Ideal for use in the chemical industry
  • Flock-lined inside
Double wall thickness: 0.60 mm
EN 388 (2111), EN 374 (KLA), EN 374-2
Certifikace: EN 388 (2111X), EN 374-1:2016 Type A (AJKLMN), EN 374-5:2016
Food contact approved
Balení: Individually packaged in poly bags.