Winter Gloves, Nitras® Winter Blocker 1603W
Supplier: AS ARBEITSSCHUTZ
1603W8 1603W11
112-2415EA 4.2 EUR
112-2415 112-2416
Winter Gloves, Nitras® Winter Blocker 1603W
Gloves
Seamless cotton terry loop gloves with latex coating on palm and fingertips.
  • knit waistband
  • Maximum comfort, maximum dexterity
  • Shrink-roughened coating for extreme grip
  • Signal colors for better visibility
EN 388 2232, EN 511 11
Certifikace: EN 388 (2241X), EN 511 (02X); Cat. II