Chemical resistant gloves, Nitras® 3470 Dual Barrier
Supplier: AS ARBEITSSCHUTZ
34707 34708 34709 347010 347011
112-2615EA 3.8 EUR
112-2615 112-2616 112-2617 112-2618 112-2619
Chemical resistant gloves, Nitras® 3470 Dual Barrier
Heavy, flock-lined bi-colour gloves made from neoprene with latex sleeves.
  • Very flexible, very good dexterity
  • Ideal for use in the chemical industry
Certifikace: EN 388 (2121X), EN ISO 374-1:2016 Type B (AKL), EN ISO 374-5:2016
Food contact approved
Balení: Individually packaged in poly bags.