Cut resistant gloves, Nitras® TAEKI5® 6705
Supplier: AS ARBEITSSCHUTZ
6705S 6705M 67052XL 67053XL 6705L 6705XL
ASAS6705SEA 91 EUR
ASAS6705S ASAS6705M ASAS67052XL ASAS67053XL ASAS6705L ASAS6705XL
Cut resistant gloves, Nitras® TAEKI5® 6705
Gloves
Knitted glove made of TAEKI5® special cut protection fiber knit (13 mesh), with black polyurethane coating on palm and fingertips.
  • Polyurethane coating for high suppleness and medium grease and oil resistance
  • Extremely high cut resistance
  • Seamless processing
  • Knitted cuff for an optimal finish on the wrist
Ideal for cutting work in the metal and automotive industries where good touch is also required.
Certifikace: EN 388 (4442C), EN 407 (X1XXXX); Cat. II