Cut resistant gloves, Nitras® TAEKI5® 6720
Supplier: AS ARBEITSSCHUTZ
6720S 6720M 6720L 6720XL 67202XL 67203XL 6720/L 6720/M 6720/S 6720/XL 6720/XXL
ASAS6720SEA 92.2 EUR
ASAS6720S ASAS6720M 112-2631 112-2632 112-2633 112-2634 ASAS6720/L ASAS6720/M ASAS6720/S ASAS6720/XL ASAS6720/XXL
Cut resistant gloves, Nitras® TAEKI5® 6720
Gloves
Knitted glove made of TAEKI5® special cut protection fiber knit (13 mesh), with black latex coating on palm and fingertips.
  • Latex coating for good grip and high acid resistance
  • Good abrasion and high cut resistance, heat resistant
  • Seamless processing
Ideal for cutting work in the metal and automotive industries where good touch is also required.
Farbe: grau
EN 388 4542
Certifikace: EN 388 (3443C), EN 407 (X2XXXX); Cat. II