General purpose gloves, Maxifoam
Supplier: Trenka
112-2654EA 5.9 EUR
112-2654 112-2653 112-2650 112-2651
General purpose gloves, Maxifoam
Gloves
Flexible, lightweight and seamless knitted nylon glove (13 gauge) with knit wrist. Partially dipped with nitrile foam.
  • Very comfortable to wear
  • Extremely sure-grip, abrasion-resistant
Very good when working with oily and greasy materials.
Certifikace: EN 388 (4131)