Industrial gloves, StrongHand® Handan 0613
Supplier: FELDTMANN HELMUT
06137 06138 061310 061311 06139
112-2280EA 0 EUR
112-2280 112-2281 112-2283 FELD061311 FELD06139
Industrial gloves, StrongHand® Handan 0613
Gloves
Seamless fine knit gloves with nitrile foam inner wall coating with nitrile nubs for better grip. The carrier fabric consists of 90% nylon (polyamide) and 10% Lycra® (spandex).
  • Excellent fit, high wearing comfort
  • Safe grip even with smooth objects
  • Very good tactile sensation
  • Good resistance to oils and greases, liquid-repellent
  • Paired in a polybag
Ideal for handling small objects, for workshops, assembly work and precision mechanics.
EN 388 4121
Certifikace: EN 388 (4121X), EN 407 (X1XXXX)