Nomex Terrycloth heat protection glove
Supplier: Trenka
112-2660EA 32.2 EUR
112-2660
Nomex Terrycloth heat protection glove
Gloves
Seamless heat protection glove with high insulation. Made of 95% Nomex® III and 5% Kevlar®.
  • Pleasant to wear; Very good fit
  • Permanently heat-resistant up to approx. 250 ° C
Ideal for handling hot parts, in bakeries, and in the glass and metal industry.
Total length: 270 mm
Certifikace: EN 388 (2241), EN 407 (32xx1x)