Jacket with waistband, flame retardant, Trenkatex® by filia ASF NOMEX® III
Supplier: Rentex
TrenkaTex®
RENT129458EA 278 EUR
RENT129458 RENT129448 RENT129460 RENT129450 RENT129452 RENT129454 RENT129456
Jacket with waistband, flame retardant, Trenkatex® by filia ASF NOMEX® III
Workwear
Chemical repellent, flame retardant and anti-static jacket made from 94% Nomex® III, 5% ParaAramid and 1% metal.
  • Increased wearer comfort due to lightweight fabric
  • Two breast pockets with press stud, one of which is a mobile phone pocket
  • One breast pocket inside, two concealed side pockets, two large inner pockets
  • Concealed press stud panel, decorative seams in combination colour
  • Adjustable cuffs
Weight: 260 g/m²
EN 531 A, B1, C1; EN 533; EN 470-1; EN 1149-1
Certifikace: EN 531 A, B1, C1; EN 533; EN 470-1; EN 1149-1