Supplier: RENTEX
129558 129548 129550 129552 129554 129556
RENT129558EA 0 EUR
RENT129558 RENT129548 RENT129550 RENT129552 RENT129554 RENT129556
Workwear
Chemical resistant, flame retardant trousers made from 94% Nomex® III, 5% Para-Aramid and 1% metal. The material is also permanently antistatic. Increased wearing comfort due to light fabric.
  • Waistband with belt loops
  • Two slanted pockets on the front, two jeans pockets with flaps on the back, ruler pocket
  • Decorative seams in combination colour
  • Fly with metal zip, covered on the inside
Weight: 260 g/m²
Certifikace: EN 531 A, B1, C1; EN 533; EN 470-1; EN 1149-1