Jacket, Trenkatex® Cotton-Tech
Supplier: RENTEX
115344 115346 115348 115350 115352 115354 115356 115358 115360 115362 121256 25540 25542 25544 25546 25548 25550 25552 25554 25556 25558 25560 25562 25564 25566 121244 121246 121248 121250 121252 121254 121258 121260 121262 121264 25538
113-05593EA 0 EUR
113-05593 113-05594 113-05595 113-05596 113-05597 113-05598 113-05599 113-05600 113-05601 113-05602 113-5535 113-06615 113-06616 113-2083 113-2084 113-2086 113-2087 113-2088 113-2089 113-2092 113-2093 113-2094 113-06617 113-06618 113-06619 113-5529 113-5530 113-5531 113-5532 113-5533 113-5534 113-5536 113-5537 113-5538 113-5539 113-06614
Jacket, Trenkatex® Cotton-Tech
Workwear
Jacket with waistband made from 100% cotton, mercerised and indanthrene dyed, with Sanfor®.
  • Breast pockets with flaps
  • Cuffs can be adjusted with press studs
  • Concealed press stud panel
  • Washable at 95 °C
Weight: Approx. 320 g/m²
Informace pro objednávání: Available in many fabric qualities, weights, special colours and also ladies' sizes (34 – 52) on request.