Overalls, Fristads® PR54-820, Design B, black
Supplier: Fristads Kansas Group
Fristads®
KWINPR54820B905462EA 0 EUR
KWINPR54820B905462 KWINPR54820B905860 KWINPR54820B905862 KWINPR54820B901548 KWINPR54820B901546 KWINPR54820B901544 KWINPR54820B901662 KWINPR54820B901660 KWINPR54820B901262 KWINPR54820B901260 KWINPR54820B905460 KWINPR54820B902062 KWINPR54820B905452 KWINPR54820B902060 KWINPR54820B905454 KWINPR54820B905850 KWINPR54820B905456 KWINPR54820B905852 KWINPR54820B905854 KWINPR54820B905458 KWINPR54820B905856 KWINPR54820B905858 KWINPR54820B901658 KWINPR54820B901656 KWINPR54820B901258 KWINPR54820B901654 KWINPR54820B901652 KWINPR54820B908160 KWINPR54820B901256 KWINPR54820B901650 KWINPR54820B901254 KWINPR54820B908162 KWINPR54820B901252 KWINPR54820B901250 KWINPR54820B905450 KWINPR54820B908158 KWINPR54820B902050 KWINPR54820B905444 KWINPR54820B905446 KWINPR54820B905844 KWINPR54820B905448 KWINPR54820B905846 KWINPR54820B905848 KWINPR54820B901648 KWINPR54820B901646 KWINPR54820B901248 KWINPR54820B901644 KWINPR54820B901246 KWINPR54820B901244 KWINPR54820B908150 KWINPR54820B902058 KWINPR54820B908152 KWINPR54820B902056 KWINPR54820B908154 KWINPR54820B902054 KWINPR54820B908156 KWINPR54820B902052 KWINPR54820B902160 KWINPR54820B908146 KWINPR54820B908148 KWINPR54820B902048 KWINPR54820B902046 KWINPR54820B902044 KWINPR54820B902162 KWINPR54820B908144 KWINPR54820B902150 KWINPR54820B909060 KWINPR54820B902158 KWINPR54820B907160 KWINPR54820B902156 KWINPR54820B909062 KWINPR54820B907162 KWINPR54820B902154 KWINPR54820B907560 KWINPR54820B902152 KWINPR54820B907562 KWINPR54820B907156 KWINPR54820B907552 KWINPR54820B907158 KWINPR54820B907554 KWINPR54820B907556 KWINPR54820B907558 KWINPR54820B909050 KWINPR54820B902148 KWINPR54820B902146 KWINPR54820B909054 KWINPR54820B909052 KWINPR54820B908360 KWINPR54820B907150 KWINPR54820B902144 KWINPR54820B909058 KWINPR54820B907152 KWINPR54820B908362 KWINPR54820B907154 KWINPR54820B907550 KWINPR54820B909056 KWINPR54820B908356 KWINPR54820B907146 KWINPR54820B909048 KWINPR54820B907544 KWINPR54820B908358 KWINPR54820B907148 KWINPR54820B907546 KWINPR54820B907548 KWINPR54820B908350 KWINPR54820B901562 KWINPR54820B909046 KWINPR54820B908352 KWINPR54820B901560 KWINPR54820B909044 KWINPR54820B908354 KWINPR54820B907144 KWINPR54820B908344 KWINPR54820B908346 KWINPR54820B908348 KWINPR54820B901558 KWINPR54820B901556 KWINPR54820B901554 KWINPR54820B901552 KWINPR54820B901550
Overalls, Fristads® PR54-820, Design B, black
Workwear
Overalls made from 65% polyester, 35% cotton, with soft nap on reverse side.
  • Zip, action back pleat
  • Two chest pockets with flap,
  • Two front pockets, two back pockets
  • Folding-ruler pocket, leg pocket with flap, sleeve pocket, mobile phone pocket, pen pocket
  • Adjustable elasticated back, adjustable cuffs
Weight: 260 g/m²
Basic colour: Black
Choice of many different accent colours (indicated in the ordering table)