Safety boots, Wellingtons, Euromaster
Supplier: FELDTMANN HELMUT KG
3536237 3536138 3536239 3536240 3536241 3536244 3536245 3536246 3536247 3536248 3536242 3536243
114-3480EA 35.6 EUR
114-3480 114-3481 114-3482 114-3483 114-3484 114-3487 114-3488 114-3489 114-3490 114-3491 114-3485 114-3486
Safety boots, Wellingtons, Euromaster
Shoes and Boots
Antistatic S5 Wellingtons made from PVC upper and a PVC/nitrile sole conditionally oil, fuel, fat and acid resistant.
  • Provides good flexibility also at low temperatures
  • Reinforcing ribs on shaft
  • Can be shortened at 2 heights
  • Protective steel toe cap (200 J)
  • Corrosion proof puncture resistant sole
Weight: 2,1 kg/pair
Shaft height: 38 cm
Shaft colour: Yellow
Sole colour: Black