Safety ankle boots, lace-up, Anatomic Bau® 845 XP
Supplier: ATLAS - the shoe company
06544 06547 06548 06536 06537 06538 06539 06540 06541 06542 06543 06545 06546 06549
114-1306EA 0 EUR
114-1306 114-1309 114-1310 114-1298 114-1299 114-1300 114-1301 114-1302 114-1303 114-1304 114-1305 114-1307 114-1308 114-1311
Safety ankle boots, lace-up, Anatomic Bau® 845 XP
Shoes and Boots
Safety ankle boot made from waterproof leather, woven fur lining and a cold isolating ICE AGE outdoor® sole (4,5 mm rough tread profile, oil, acid and petrol resistant).
  • Isolating supporting surface and warm woven fur lining
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Puncture resistant, metal-free XP® midsole
  • Large stud profile
  • Reinforced toe protection
Colour: Black/blue
Certifikace: DGUV 112-191; EN ISO 20345; S3, CI, SRC