Occupational shoes, lace-up, Airpower® C7
Supplier: HAIX SCHUHE
1003024.5 1003025 1003026 1003026.5 1003027.5 1003028 1003029 1003029.5 10030210.5 10030211 10030212 10030213 10030213.5 10030214.5
114-1610EA 181 EUR
114-1610 114-1611 114-1612 114-1613 114-1614 114-1615 114-1616 114-1617 114-1604 114-1605 114-1606 114-1607 114-1608 114-1609
Occupational shoes, lace-up, Airpower® C7
Shoes and Boots
Sporty service and leisure shoe made of 1.8 - 2.0 mm waterproof leather and Gore-Tex® lining.
The TPU/PU profile sole with a street profile has high cushioning properties and cold insulation of the sole complex. It is oil and petrol resistant and antistatic.
  • Waterproof and breathable thanks to the integrated Gore-Tex® membrane
  • Insole supports the natural arch of the midfoot; Foot malpositions are prevented
  • Orthopedic properties ensure pressure relief, cushioning, correction of foot position, body straightening and muscle balance
  • Damping with damping wedge
  • Sun reflection effect reduces heating of the upper leather
Colour: Black