Occupational boots, lace-up, Tibet
Supplier: HAIX SCHUHE
2010025 2010026 2010026.5 2010027.5 2010028 2010029 2010029.5 20100210.5 20100211 20100212
HAIX2010025EA 0 EUR
HAIX2010025 HAIX2010026 HAIX2010026.5 HAIX2010027.5 HAIX2010028 HAIX2010029 HAIX2010029.5 HAIX20100210.5 HAIX20100211 HAIX20100212
Occupational boots, lace-up, Tibet
Shoes and Boots
Lace-up work boots made of 2.5-2.7 mm waterproof leather and Gore-Tex® lining. The rubber/PU sole with road/terrain profile is abrasion-resistant and non-slip, light and provides excellent insulation against cold and heat.
  • Waterproof and breathable thanks to Gore-Tex® lining
  • Breathable, moisture is transported to the ventilation openings on the shaft and tongue
  • Sun reflection effect reduces heating of the upper leather
  • Increased impact absorption and isolation in the heel area
Colour: Black
Shaft height: 23 cm
Width: 11