Safety shoes, lace-up, Airpower® R22 low
Supplier: HAIX SCHUHE
60780645 60780648 60780636 60780637 60780638 60780639 60780640 60780641 60780642 60780643 60780644 60780647 60780649 60780650
114-1895EA 0 EUR
114-1895 114-1898 114-1886 114-1887 114-1888 114-1889 114-1890 114-1891 114-1892 114-1893 114-1894 114-1897 114-1899 114-1900
Safety shoes, lace-up, Airpower® R22 low
Shoes and Boots
Lace-up safety shoe made of waterproof leather (2.0 - 2.2 mm thick) with leather lining, replaceable and anatomically shaped insole (washable). The rubber/PU outsole with a sporty street and terrain profile has high cushioning properties and contact heat resistance, ensures heat and cold insulation of the sole complex and is oil and petrol resistant and antistatic.
  • Insole supports the natural arch of the midfoot; Foot malpositions are prevented
  • Increased impact absorption and isolation in the heel area
  • Orthopedic properties ensure pressure relief, cushioning, correction of foot position, body straightening and muscle balance
  • Sun reflection effect reduces heating of the upper leather
  • Extra wide and high steel toe cap
  • Penetration-resistant midsole
Colour: Black
width: 11