ESD occupational shoes, clogs, Tiefbett Leder, Munich, 11320, 11420
Supplier: SCHURR SCHUHVERTRIEB
1.1420.01.35 1.1420.01.36 1.1420.01.37 1.1420.01.38 1.1420.01.39 1.1420.01.40 1.1320.01.1441 1.1320.01.1442 1.1320.01.1444 1.1320.01.1445 1.1320.01.1447 1.1320.01.1448
SCRR1.1420.01.35EA 74.5 EUR
SCRR1.1420.01.35 SCRR1.1420.01.36 SCRR1.1420.01.37 SCRR1.1420.01.38 SCRR1.1420.01.39 SCRR1.1420.01.40 SCRR1.1320.01.1441 SCRR1.1320.01.1442 SCRR1.1320.01.1444 SCRR1.1320.01.1445 SCRR1.1320.01.1447 SCRR1.1320.01.1448
ESD occupational shoes, clogs, Tiefbett Leder, Munich, 11320, 11420
Shoes and Boots
Deeply moulded occupational shoes made from full grain leather and an anti-slip and anti-static "A4" nitrile rubber sole; with adjustable heel strap.
  • Anatomically shaped stabilising integrated insole made from cork/latex
  • Insole features cushioned heel, ankle support and toe bar
  • Shock absorber heel
For ESD and kitchen use.
Colour: White
Width: 9
Weight: 300 g
Certifikace: EN 61340 (ESD); EN ISO 20347:2012; OB, SRC