Ergo-Med® 360 ESD Safety Shoes, Velcro® Closure
Supplier: ATLAS - the shoe company
87738/W12 87736/W12 87737/W12 87739/W12 87740/W12 87741/W12 87742/W12 87743/W12 87744/W12 87745/W12 87746/W12 87747/W12 87748/W12 87749/W12 87750/W12
ATLA87738/W12EA 0 EUR
ATLA87738/W12 ATLA87736/W12 ATLA87737/W12 ATLA87739/W12 ATLA87740/W12 ATLA87741/W12 ATLA87742/W12 ATLA87743/W12 ATLA87744/W12 ATLA87745/W12 ATLA87746/W12 ATLA87747/W12 ATLA87748/W12 ATLA87749/W12 ATLA87750/W12
Ergo-Med® 360 ESD Safety Shoes, Velcro® Closure
Shoes and Boots
Extra wide ESD safety sandals feature Velcro® fastening and reflectors in special width 14 and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Moisture control and breathability thanks to Clima-stream® technology
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Carbon fibre reinforced toe protection
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Width: 14
Colour: Black
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S1