Lithium chloride 8 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
L7026-100ML L7026-1L L7026-500ML
SIALL7026-100MLEA 120 EUR
SIALL7026-100ML SIALL7026-1L SIALL7026-500ML
Lithium chloride 8 mol/l in aqueous solution, Sigma-Aldrich®
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382
Melting Pt: 614
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550