N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-[Dec-1-yl(dimethyl)ammonio]propane-1-sulphonate, 3-(Decyldimethylazaniumyl)propane-1-sulphonate
M-Clarity™ quality level = MQ200
D4266-25G D4266-5G
SIALD4266-25GEA 324 EUR
SIALD4266-25G SIALD4266-5G
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-10, SB-10), Sigma-Aldrich®
N-Decyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-10, SB-10)
Formula: C₁₅H₃₃NO₃S
MW: 307.4924 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036908
CAS Number: 15163-36-7