N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt
M-Clarity™ quality level = MQ200
40772-250G 40772-50G
SIAL40772-250GEA 263 EUR
SIAL40772-250G SIAL40772-50G
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14), Sigma-Aldrich®
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-14, SB-14)
Formula: C₁₉H₄₁NO₃S
MW: 363.5988 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036910
CAS Number: 14933-09-6
EINECS: 239-003-9