AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Supplier: PanReac AppliChem
Warning
Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
A1421.0500 A1421.0001
A1421.0500EA 254 EUR
A1421.0500 A1421.0001
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)
Formula: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00132962
CAS Number: 30827-99-7
UN: 3261
ADR: 8,II