AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)

Supplier: ROCHE


M-Clarity™ quality level = MQ100

ROCH11429868001EA 144 EUR
ROCH11429868001 ROCH1429868
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formula: C₈H₁₁ClFNO₂S
MW: 239,69 g/mol
MDL Number: MFCD00132962
CAS Number: 30827-99-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR