Guanosine, Ultra Pure Grade for biochemistry

Supplier: VWR Chemicals
Danger

0193-100GEA 292 EUR
0193-100G
Guanosine, Ultra Pure Grade for biochemistry
Guanosine
Formula: C₁₀H₁₃N₅O₅
MW: 283,24 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00010182
CAS Number: 118-00-3

Order Now

Specification Test Results

Em (252.5 nm, 0.01 M PO₄ buffer) 12500 – 15000

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR