Guanosine

Supplier: Roth Carl

ROTH0715.1EA 27.3 EUR
ROTH0715.1
Guanosine
Guanosine
Formula: C₁₀H₁₃N₅O₅
MW: 283,24 g/mol
MDL Number: MFCD00010182
CAS Number: 118-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR