Capillary columns, Elite-17 for autosystem
Supplier: PerkinElmer
N9316115 N9316123 N9316131 N9316137 N9316121 N9316122 N9316128 N9316135 N9316136
PERCN9316115EA 1620 EUR
PERCN9316115 PERCN9316123 PERCN9316131 PERCN9316137 PERCN9316121 PERCN9316122 PERCN9316128 PERCN9316135 PERCN9316136
Capillary columns, Elite-17 for autosystem
Chromatography Columns
Elite-17 (50% Phenyl) Methylpolysiloxane series capillary columns are ideal for confirmation analysis and are inert for active compounds.