Lotion soaps, Plum No. 14
Supplier: Plum Safety
1403 1413
PLUM1403EA 18.5 EUR
PLUM1403 115-1370
Lotion soaps, Plum No. 14
Skincare Soaps and Skin Cleansers
Perfumed luxery soap for normal daily hand washing in toilets, offices or production facilities.
  • Without preservatives
  • For dispenser system MP2000
Upozornení: *The product(s) on this page are outside the scope of the EU Directive 2019/904