2-Propyl-1-heptanol ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Propylheptan-1-ol, 2-n-Propyl-1-heptanol
H50340.03 H50340.06
H50340.03EA 244 EUR
H50340.03 H50340.06
2-Propyl-1-heptanol ≥98%
2-Propyl-1-heptanol
Purity: 98% CAS No.: 10042-59-8 Molecular Formula: C10H22O Formula Weight: 158.28 MDL No.: MFCD00046768
Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046768
CAS Number: 10042-59-8
EINECS: 233-126-1