Glutaryl dichloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Glutaryl chloride, Glutaryl dichloride, Glutaroyl dichloride
M-Clarity™ quality level = MQ100
G4608-100G
G4608-25G
SIALG4608-100GEA
226
EUR
SIALG4608-100G
SIALG4608-25G
Glutaryl dichloride, Sigma-Aldrich®
Glutaryl dichloride
Formula:
C₅H₆Cl₂O₂ MW: 169.0059 g/mol |
MDL Number:
MFCD00000755 CAS Number: 2873-74-7 EINECS: 220-711-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...