Nonidet® P 40 Substitute (NP-40)
Supplier: G Bioscienes
Warning
786-511 786-512
786-511EA 445 EUR
786-511 786-512
Nonidet® P 40 Substitute (NP-40)
Nonidet® P 40 Substitute (NP-40)
Storage Temperature: Ambient
MDL Number: MFCD00132851
CAS Number: 9016-45-9
EINECS: 500-024-6