Hydroxypropyl cellulose, M.W. 100000, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
191884-100G 191884-250G 191884-5G
SIAL191884-100GEA 177 EUR
SIAL191884-100G SIAL191884-250G SIAL191884-5G
Hydroxypropyl cellulose, M.W. 100000, Sigma-Aldrich®
Hydroxypropyl cellulose
MDL Number: MFCD00132688
CAS Number: 9004-64-2