Hydroxypropyl cellulose, M.W. 1000000, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

191906-100G 191906-250G
SIAL191906-100GEA 150 EUR
SIAL191906-100G SIAL191906-250G
Hydroxypropyl cellulose, M.W. 1000000, Sigma-Aldrich®
Hydroxypropyl cellulose
MDL Number: MFCD00132688
CAS Number: 9004-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR