Hydroxypropyl cellulose, M.W. 370000, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

191892-100G 191892-250G 191892-5G
SIAL191892-100GEA 152 EUR
SIAL191892-100G SIAL191892-250G SIAL191892-5G
Hydroxypropyl cellulose, M.W. 370000, Sigma-Aldrich®
Hydroxypropyl cellulose
MDL Number: MFCD00132688
CAS Number: 9004-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR