Hydroxypropyl methylcellulose (Hypromellose), M.W. 86,000, 4000 cP (2% H₂O)
Supplier: Thermo Fisher Scientific
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
244021000 244022500 244020010
ACRO244021000EA 155 EUR
ACRO244021000 ACRO244022500 ACRO244020010
Hydroxypropyl methylcellulose (Hypromellose), M.W. 86,000, 4000 cP (2% H₂O)
Hydroxypropyl methyl cellulose (Hypromellose)
Formula: C₃H₈O₂·xCH₄O
Density: 1.39 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00131360
CAS Number: 9004-65-3
Order Now
Specification Test Results
Appearance White to cream Powder
Infrared spectrum Conforms
Loss on drying ≤10 %