Hydroxypropyl methylcellulose (Hypromellose), M.W. ~10,000, 6 cP, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methocel K100, Hydroxypropyl methyl cellulose
M-Clarity™ quality level = MQ100
423238-100G 423238-25G
SIAL423238-100GEA 130 EUR
SIAL423238-100G SIAL423238-25G
Hydroxypropyl methylcellulose (Hypromellose), M.W. ~10,000, 6 cP, Sigma-Aldrich®
Hydroxypropyl methyl cellulose (Hypromellose)
Formula: C₃H₈O₂·xCH₄O
MDL Number: MFCD00131360
CAS Number: 9004-65-3