1,3-Diisopropylimidazolium chloride ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1,3-Di-i-propylimidazolium chloride, 1,3-Diisopropyl-1H-imidazol-3-ium chloride
H27798.03 H27798.06
H27798.03EA 70.6 EUR
H27798.03 H27798.06
1,3-Diisopropylimidazolium chloride ≥97%
1,3-Diisopropylimidazolium chloride
Purity: 97+% CAS No.: 139143-09-2 Molecular Formula: C9H17ClN2 Formula Weight: 188.69 Melting Point: 182-186° MDL No.: MFCD03840577
Formula: C₉H₁₇ClN₂
MW: 188.70 g/mol
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD03840577
CAS Number: 139143-09-2