Tiglic acid
Supplier: Thermo Scientific
Synonyms: (E)-2-Methylbut-2-enoic acid, trans-2,3-Dimethylacrylic acid
SB00765.1 SB00765.10 SB00765.50
MAYBSB00765.1EA 0 EUR
MAYBSB00765.1 MAYBSB00765.10 MAYBSB00765.50
Tiglic acid
Tiglic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00066864
CAS Number: 80-59-1
EINECS: 201-295-0
UN: 3261
ADR: 8,III