N-Acetyl-β-D-glucosamine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

M-Clarity™ quality level = MQ200

A8625-10MG A8625-5G
SIALA8625-10MGEA 35.2 EUR
SIALA8625-10MG SIALA8625-5G
N-Acetyl-β-D-glucosamine, Sigma-Aldrich®
N-Acetyl-β-D-glucosamine
Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00136044
CAS Number: 7512-17-6
EINECS: 231-368-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR