2-Deoxy-D-glucose, Sigma-Aldrich®

Supplier: Merck

Synonyms: (3R,4S,5R)-3,4,5,6-Tetrahydroxyhexanal

MQ200

D3179-1G D3179-250MG D3179-5G
SIALD3179-1GEA 274 EUR
SIALD3179-1G SIALD3179-250MG SIALD3179-5G
2-Deoxy-D-glucose, Sigma-Aldrich®
2-Deoxy-D-glucose
Formula: C₆H₁₂O₅
MW: 164.16 g/mol
MDL Number: MFCD00151328
CAS Number: 154-17-6
EINECS: 205-823-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR